Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FastAP Thermosensitive Alkaline Phosphatase Promotion

Catalyze the removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins using this thermosensitive alkaline phosphatase .

Manufacturer:  Thermo Scientific™ EF0652

 View more versions of this product

Catalog No. FEREF0652PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

FastAP Thermosensitive Alkaline Phosphatase catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA, and nucleotides. This enzyme also removes phosphate groups from proteins.

FastAP is a novel alkaline phosphatase, which is active in all Thermo Scientific restriction enzyme buffers as well as in PCR buffers. It dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs) in 10 minutes at 37°C. The enzyme is inactivated in 5 minutes at 75°C . Therefore, removal of alkaline phosphatase is not required prior to ligation.

Highlights

  • Recombinant enzyme
  • Fast dephosphorylation -10 minutes at 37°C
  • Fast and complete inactivation -5 minutes at 75°C
  • Simultaneous digestion and dephosphorylation of vector DNA
  • 100% active in restriction enzyme and PCR buffers
  • One protocol for all types of DNA ends:
    • 5'-overhangs
    • 3'-overhangs
    • blunt-ends
    • single nucleotides
  • PCR clean-up in conjunction with Exo I
  • Protein dephosphorylation
Specifications

Specifications

metal chelators
E.coli cells with a cloned bacterial AP gene
5 x 1000U
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

The purchase of this product allows the purchaser to use it for preparing amplified DNA fragments under a license from GE Healthcare of U.S. Patent Nos. 5,741,676 and 5,756,285 and other foreign patents.