Learn More
Thermo Scientific FastDigest Bsp1407I Promotion
Cut at T^GTACA sites with Bsp1407I restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BsrGI, BstAUI).
Manufacturer: Thermo Scientific FD0933
Description
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Highlights
- 100% activity of all FastDigest enzymes in the universal buffer
- 100% buffer compatibility with downstream applications
- Complete digestion in 5-15 minutes
- Direct loading on gels
- No star activity
- 176 FastDigest enzymes available
Specifications
FastDigest™ Bsp1407I | |
10X FastDigest Buffer and 10X FastDigest Green Buffer | |
50μL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.