Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FASTKD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550412

Catalog No. PA550412


Add to cart

Description

Description

The antibody detects endogenous levels of total FASTKD1 protein.

As a member of the FASTKD family, FASTKD1 localizes to the mitochondria to modulate their energy balance, especially under conditions of stress. Though ubiquitously expressed in all tissues, FASTKD1 appears more abundantly in skeletal muscle, heart muscle, and other tissues enriched in mitochondria. FASTKD1 has been validated as an RNA-binding protein
Specifications

Specifications

FASTKD1
Polyclonal
Unconjugated
FASTKD1
Liquid
FASTKD1
Fusion protein corresponding to residues near the C terminal of human FAST kinase domains 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.5 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q53R41
KIAA1800
Rabbit
IgG
100 μL
-20°C
Primary
79675
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit