Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FASTKD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550413
Description
The antibody detects endogenous levels of total FASTKD5 protein.
FASTKD5 Polyclonal antibody specifically detects FASTKD5 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FASTKD5 | |
Polyclonal | |
Unconjugated | |
FASTKD5 | |
C78212; dJ1187M17.5; FAST kinase domain-containing protein 5; FAST kinase domain-containing protein 5, mitochondrial; FAST kinase domains 5; FAST kinase domains 5, pseudogene 1; Fastkd5; Fastkd5-ps1; KIAA1792; mKIAA1792; RGD1563839 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60493 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7L8L6 | |
FASTKD5 | |
Fusion protein corresponding to a region derived from internal residues of human FAST kinase domains 5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction