Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fasudil hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Fasudil hydrochloride is a cyclic nucleotide-dependent protein kinase inhibitor and Rho-associated kinase inhibitor (IC50 = 10.7 μM). Fasudil suppresses MMP-2 expression and induces apoptosis in glioblastoma cells in vivo. Fasudil is a Ca2+ antagonist, vasodilator and inhibits proliferation of vascular smooth muscle cells. Fasudil binds to α-synuclein to reduce aggregate formation in cellular models of Parkinson's disease, also displays neuroprotective properties and increases survival of dopaminergic neurons in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 327.83 |
Percent Purity | >98% |
Chemical Name or Material | Fasudil hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction