Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

FAT1 Mouse anti-Drosophila, Clone: FAT1-3D7/1, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen FAT1
Clone FAT1-3D7/1
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121886
View Documents
Novus Biologicals
NBP30720620UG
20 μg
Each for $138.37
N/A
NB121885
View Documents
Novus Biologicals
NBP30720610UG
100 μg
Each for $352.89
N/A
Description

Description

FAT1 Monoclonal antibody specifically detects FAT1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications

Specifications

FAT1
0.2 mg/ml
Western Blot, ELISA, Immunofluorescence
Purified
RUO
Drosophila
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Cytoplasmic domain of Drosophila FAT1 protein.
Monoclonal
Protein L purified
FAT1-3D7/1
Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Signal Transduction
10 mM PBS with 0.05% BSA
2195
IgM Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit