Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAT1 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232275H

Catalog No. NBP232275H


Add to cart

Description

Description

FAT1 Polyclonal antibody specifically detects FAT1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FAT1
HRP
PBS with No Preservative
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Rabbit
IgG
100 ul
Signal Transduction
Primary
2195
Western Blot
Western Blot
Affinity Purified
FAT1
The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195).
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only