Learn More
FATP5/SLC27A5 Mouse anti-Human, Mouse, DyLight 350, Clone: 4B11C10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237413UV
Catalog No. NBP237413UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FATP5/SLC27A5
4B11C10
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SLC27A5
Purified recombinant fragment of human SLC27A5 (AA: 508-570) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ACSBBACS, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, solute carrier family 27 (fatty acid transporter), member 5, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related
Mouse
IgG1
0.1 ml
Primary
10998