Learn More
Fatty Acid Synthase/FASN Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: 237, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243811UV
Catalog No. NBP243811UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Fatty Acid Synthase/FASN
237
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
FASN
Recombinant protein encompassing a sequence within the N-terminus region of human Fatty Acid Synthase. The exact sequence is proprietary.
Protein G purified
Apoptosis, Cancer, Diabetes Research, Lipid and Metabolism, Neuroendocrinology
Primary
2194
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat