Learn More
Braintree ScientificSupplier Diversity Partner FB-MEDIUM RATS RESTRAINER DFS Item
Manufacturer:  Braintree ScientificSupplier Diversity Partner FBM
Individual semicylindrical plastic restrainers has an open slot on the bottom (for intraperitoneal injections), a hole on the top (for catheters exiting the back of the neck), and a slotted tail piece (for intravenous injections).
Catalog No. NC0180926
Due to product restrictions, please to purchase or view availability for this product.