Learn More
Fisherbrand Isotemp General Purpose Laboratory Refrigerators, Solid-Stainless Steel Door
Ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, and proteins. Fisherbrand Isotemp General Purpose Laboratory Refrigerators are designed for your everyday laboratory needs.
Description
- Whisper-quiet operation
- ENERGY STAR certified
- Temperature, door ajar, and power failure (with battery back-up) alarms
- Green, natural refrigerants
- Adaptive defrost
- Glass (double pane) or solid door configurations
- Choice of white painted interior and exterior, or stainless steel interior and gray exterior
- Digital controller
- Rear access port
- Remote alarm contacts
- Keyed door lock(s)
- LED interior lighting
- Low heat output
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Solid Stainless Steel | |
2°C to 10°C | |
27 cu. ft. | |
115 V 60 Hz | |
NEMA 5-15 P | |
28.5 x 27 x 61.75 in. (724 x 686 x 1569 mm) | |
175 kg | |
Painted White Steel | |
45 kg | |
2 years parts and labor, plus 3 year compressor warranty | |
5°C | |
1 Rear access port |
4 Full | |
cULus, ENERGYSTAR | |
764 L | |
3 A | |
32 x 30.75 x 81 in.(813 x 781 x 2057 mm) | |
386 lb. | |
R290 | |
100 lb. | |
Key Lock | |
White | |
RPSA | |
Microprocessor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.