Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBL Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

$346.00

Specifications

Antigen FBL
Host Species Rabbit
Isotype IgG
Target Species Human
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-033-357 View Documents Abnova Corporation
PAB30069
100 uL Each for $346.00
Add to cart
 
89-035-702 View Documents Abnova Corporation
PAB30209
40 ug Each for $346.00
Add to cart
 
Description

Description

Rabbit polyclonal antibody raised against synthetic peptide of human FBL.

  • High specificity
  • High reactivity
Specifications

Specifications

FBL
IgG
Western Blot
Unconjugated
Store at -20°C on dry atmosphere, lyophilized antibodies are stable.
After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Rabbit
Human
RUO
Lyophilized
Primary
Lyophilized from PBS, pH 7.4 (0.02% sodium azide).

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit