Learn More
Alfa Aesar™ FBPase-1 Inhibitor
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
Manufacturer:  Alfa Aesar™ J65298MF
 View more versions of this product
Catalog No. AAJ65298MF
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
50mg
2° to 8°C
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
C13H7Cl3N2O3S
F16BPase Inhibitor
Soluble in DMSO
Off-white
Solid
883973-99-7
377.60g/mol
Light Sensitive
Signal Transduction Reagent