Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBPase-1 Inhibitor

A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1

$121.30 - $502.50

Specifications

CAS 883973-99-7
Formula Weight 377.60g/mol
Synonym F16BPase Inhibitor
Storage Requirements 2° to 8°C
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ65298MF View Documents Alfa Aesar™
J65298MF
50mg Each for $502.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ65298MA View Documents Alfa Aesar™
J65298MA
10mg Each for $121.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

883973-99-7
F16BPase Inhibitor
Solid
A cell-permeable benzoxazolo-sulfonamide compound that inhibits hFBPase-1
Signal Transduction Reagent
377.60g/mol
2° to 8°C
Light sensitive
Off-white
Soluble in DMSO
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit