Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBPase 1 Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF7285SP
This item is not returnable.
View return policy
Description
FBPase 1 Polyclonal antibody AF7285SP specifically detects FBPase 1 in Human, Mouse, Rat samples. It is validated for Western BlottingSpecifications
FBPase 1 | |
LYOPH | |
Western Blot 0.2 ug/mL | |
P09467 | |
D-fructose-1,6-bisphosphate 1-phosphohydrolase 1, EC 3.1.3, EC 3.1.3.11, FBP, FBP1, FBPase 1, fructose-1,6-bisphosphatase 1, fructose-bisphosphatase 1, growth-inhibiting protein 17 | |
Sheep | |
IgG | |
25 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human, mouse, rat FBPase 1 in Western blots and detects human FBPase 1 in direct ELISAs. | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
FBP1 | |
E. coli-derived recombinant human FBPase 1 Ala2-Gln338 (Arg218Lys) Accession # P09467 | |
Immunogen affinity purified | |
RUO | |
Primary | |
2203 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only