Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FBRS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560615
Description
Immunogen sequence: LRHQEKMKGD SHKLDFRNDL LPCLPGPYGA LPPGQELSHP ASLFTATGAV HAAANPFTAA PGAHGPFLSP STH Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Fibrosin is a lymphokine secreted by activated lymphocytes that induces fibroblast proliferation.
Specifications
FBRS | |
Polyclonal | |
Unconjugated | |
Fbrs | |
Fbrs; FBS; FBS1; fbsh; fibrogenic lymphokine; fibrosin; fibrosin 1; fibrosin-1; FLJ11618; probable fibrosin-1; probable fibrosin-1 long transcript protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14123, 64319, 691899 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8R089, Q9HAH7 | |
Fbrs | |
Recombinant protein corresponding to Human FBRS. Recombinant protein control fragment (Product #RP-91948). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction