Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBX09 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP155054

 View more versions of this product

Catalog No. NBP155054

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

FBX09 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blotting.
Specifications

Specifications

FBX09
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
FBXO9
Synthetic peptides corresponding to FBXO9(F-box protein 9) The peptide sequence was selected from the middle region of FBXO9. Peptide sequence PELESSQIHISVLPMEVLMYIFRWVVSSDLDLRSLEQLSLVCRGFYICAR.
52 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Yeast: 85%.
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence
Q9UK97
Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
26268
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit