Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO41 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548401

Catalog No. PA548401


Add to cart

Description

Description

Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications

Specifications

FBXO41
Polyclonal
PBS with 0.09% sodium azide
Q6NS60
F-box only protein 41, FBXO41, FBX41, KIAA1940
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
330369
Western Blot
Unconjugated
Fbxo41
Liquid
FBXO41
KLH conjugated synthetic peptide between 235-263 amino acids from the N-terminal region of human FBX41.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit