Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548821

Catalog No. PA548821

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications

Specifications

FBXL13
Polyclonal
PBS with 0.09% sodium azide
Q8NEE6
F-box/LRR-repeat protein 13, F-box and leucine-rich repeat protein 13, FBXL13, FBL13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
222235
Western Blot
Unconjugated
FBXL13
Liquid
FBXL13
KLH conjugated synthetic peptide between 683-710 amino acids from the C-terminal region of human FBXL13.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit