Learn More
FBXL13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 171391AP
Catalog No. 50-173-1400
Add to cart
Description
Description
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.
Specifications
Specifications
FBXL13
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CDU4, Q8NEE6
F box/LRR repeat protein 13, Fbl13, FBXL13
Rabbit
IgG
150 μL
-20°C
Primary
100125363, 222235, 320118
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FBXL13
Liquid
Fbxl13
FBXL13 Fusion Protein Ag10903
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat