Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXL3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP153021
Description
FBXL3 Polyclonal specifically detects FBXL3 in Human samples. It is validated for Western Blot.Specifications
FBXL3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9UKT7 | |
FBXL3 | |
Synthetic peptides corresponding to FBXL3(F-box and leucine-rich repeat protein 3) The peptide sequence was selected from the middle region of FBXL3. Peptide sequence LISTARPSFMDLPKSHFISALTVVFVNSKSLSSLKIDDTPVDDPSLKVLV. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 100%; Xenopus: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Yeast, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
FBL3AF-box and leucine-rich repeat protein 3AF-box protein Fbl3a, FBL3F-box/LRR-repeat protein 3, F-box and leucine-rich repeat protein 3, F-box/LRR-repeat protein 3A, FBXL3A | |
Rabbit | |
Affinity Purified | |
RUO | |
26224 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title