Learn More
Abnova™ FBXL5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00026234T01
Catalog No. 89-944-160
Add to cart
Specifications
Specifications
Western Blot
293T
BC030656.1
pCMV-FBXL5 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FBXL5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 76.12kDa
Transient overexpression cell lysate was tested with anti-FBXL5 antibody (Mfr. No. H00026234-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)