Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FBXL5
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093362
View Documents
Novus Biologicals
NBP304665100UL
100 μg Each for $320.71
Add to cart
 
NB093363
View Documents
Novus Biologicals
NBP30466520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

FBXL5 Polyclonal antibody specifically detects FBXL5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

FBXL5
Unconjugated
Rabbit
Human, Mouse, Rat
FBL5FBL4F-box protein FBL5, F-box and leucine-rich repeat protein 5p45SKP2-like protein, F-box protein FBL4/FBL5, FLR1F-box/LRR-repeat protein 5
Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human FBXL5 (NP_036293.1).
Polyclonal
Affinity purified
Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
26234
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit