Learn More
FBXL8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 165051AP
Catalog No. 50-173-0996
Add to cart
Description
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. It shares 78% sequence identity with the mouse protein.
Specifications
Specifications
FBXL8
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CIG9, Q96CD0
F box protein FBL8, F box/LRR repeat protein 8, FBL8, FBXL8
Rabbit
IgG
150 μL
-20°C
Primary
498941, 50788, 55336
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FBXL8
Liquid
Fbxl8
FBXL8 Fusion Protein Ag9685
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat