Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO21 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI2C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271711APCCY7

Catalog No. N271711APC7


Add to cart

Description

Description

FBXO21 Monoclonal antibody specifically detects FBXO21 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

FBXO21
OTI2C7
Flow Cytometry
Affinity Purified, Conjugated
FBXO21
Full length recombinant protein of human FBXO21 (NP_296373) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
DKFZp434G058, F-box only protein 21, F-box protein 21, FBX21FLJ90233, KIAA0875MGC26682
Mouse
IgG2a
0.1 mL
Primary
23014.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit