Learn More
Aviva Systems Biology Corp FBXO22 Antibody - middle region (ARP54860_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54860_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO22. Alternate names: FBX22, FLJ13986, MGC31799, FISTC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61347