Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO22 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549072

Catalog No. PA549072


Add to cart

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Two transcript variants encoding different isoforms exist for this gene.
Specifications

Specifications

FBXO22
Polyclonal
PBS with 0.09% sodium azide
Q78JE5
F-box only protein 22, F-box protein FBX22p44, FBXO22, FBX22
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
71999
Western Blot
Unconjugated
Fbxo22
Liquid
FBXO22
KLH conjugated synthetic peptide between 320-347 amino acids from the C-terminal region of human FBXO22.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit