Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
FBXO25 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30073
Catalog No. 89-033-361
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human FBXO25.
  • High specificity
  • High reactivity
Specifications
Specifications
FBXO25
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
NM_012173
FBX25|MGC20256|MGC51975
Rabbit
Protein A purification
RUO
Primary
26260
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human FBXO25.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
FBXO25
A synthetic peptide corresponding to C-terminus of human FBXO25.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)