Learn More
Aviva Systems Biology Corp FBXO31 Antibody - middle region (ARP53776_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53776_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXO31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXO31. Alternate names: DKFZP434B027, DKFZp434J1815, FBX14, FBXO14, FLJ22477, Fbx31, MGC15419, MGC9527, pp2386ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61125