Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO31 Mouse anti-Human, Clone: OTI1H1, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503945

Catalog No. 50-167-4721


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 , cullin , and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications

Specifications

FBXO31
Monoclonal
0.85 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5XUX0
FBX14, FBXO14, Fbx31, MRT45, pp2386
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79791
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1H1
Unconjugated
FBXO31
Liquid
FBXO31
Full length human recombit protein of human FBXO31 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit