Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO33 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549503

Catalog No. PA549503


Add to cart

Description

Description

Substrate recognition component of a SCF (SKP1-CUL1-F- box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Probably recognizes and binds to phosphorylated target proteins. Recognizes YBX1.
Specifications

Specifications

FBXO33
Polyclonal
Unconjugated
Fbxo33
Liquid
FBXO33
KLH conjugated synthetic peptide between 416-449 amino acids from human FBXO33.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Mouse
Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q8VE08
F-box only protein 33, FBXO33, FBX33
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
70611
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit