Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO42 Mouse anti-Human, Clone: OTI1H4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526418

Catalog No. PIMA526418


Add to cart

Description

Description

Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (SKP1A; MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004
Specifications

Specifications

FBXO42
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P3S6
FBXO42, Fbx42, JFK
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54455
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1H4
Unconjugated
FBXO42
Liquid
FBXO42
Human recombit protein fragment corresponding to amino acids 356-717 of FBXO42 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit