Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO47 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548173

Catalog No. PA548173


Add to cart

Description

Description

Probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation.
Specifications

Specifications

FBXO47
Polyclonal
PBS with 0.09% sodium azide
Q5MNV8, A2A6H3
F-box only protein 47, FBXO47
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
494188, 72973
Immunohistochemistry (Paraffin)
Unconjugated
FBXO47
Liquid
FBXO47
KLH conjugated synthetic peptide between 114-143 amino acids from the N-terminal region of human FBXO47.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit