Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO5 Mouse anti-Human, Clone: EMI1/1176, Abnova™

Mouse monoclonal antibody raised against partial recombinant human FBXO5.

Manufacturer:  Abnova Corporation MAB13276

Catalog No. 89-934-124


Add to cart

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is similar to xenopus early mitotic inhibitor-1 (Emi1), which is a mitotic regulator that interacts with Cdc20 and inhibits the anaphase promoting complex. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Specifications

Specifications

FBXO5
EMI1/1176
Mouse monoclonal antibody raised against partial recombinant human FBXO5.
In 1mM PBS.
EMI1/FBX5/Fbxo31
Mouse
IgG2a, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determ
Liquid
FBXO5
Recombinant protein corresponding to 203 residues around amino acids 1-250 of human FBXO5.
Protein A/G purification
RUO
Primary
26271
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit