Learn More
FBXO6 Mouse anti-Human, Clone: OTI6A4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810363
Catalog No. 50-167-0033
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class, and its C-terminal region is highly similar to that of rat NFB42 (neural F Box 42 kDa) which may be involved in the control of the cell cycle.
Specifications
Specifications
FBXO6
Monoclonal
Unconjugated
FBXO6
Lyophilized
FBXO6
Full length human recombit protein of human FBXO6 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI6A4
PBS with 8% trehalose and no preservative; pH 7.3
Q9NRD1
FBG2, FBS2, FBX6, Fbx6b
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26270