Learn More
FBXO8 Mouse anti-Human, Clone: OTI7E12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807552
Catalog No. 50-166-8331
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It contains a C-terminal amino acid sequence that bears a significant similarity with a portion of yeast Sec7p, a critical regulator of vesicular protein transport. This human protein may interact with ADP-ribosylation factor(s)(ARFs) and exhibit ARF-GEF (guanine nucleotide exchange factor) activity.
Specifications
Specifications
FBXO8
Monoclonal
Unconjugated
FBXO8
Lyophilized
FBXO8
Human recombit protein fragment corresponding to amino acids 1-231 of human FBXO8 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI7E12
PBS with 8% trehalose and no preservative; pH 7.3
Q9NRD0
DC10, FBS, FBX8
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
26269