Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FBXW10 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP179495
Description
FBXW10 Polyclonal specifically detects FBXW10 in Human samples. It is validated for Western Blot.Specifications
FBXW10 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
F-box and WD repeat domain containing 10, F-box protein, F-box/WD repeat-containing protein 10 | |
Rabbit | |
Affinity Purified | |
RUO | |
10517 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FBXW10 | |
Synthetic peptide directed towards the N terminal of human FBXW10The immunogen for this antibody is FBXW10. Peptide sequence SPEKDHSSKSATSQVYWTAKTQHTSLPLSKAPENEHLLGAASNPEEPWRN. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Crab-eating macaque: 85%. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title