Learn More
Aviva Systems Biology Corp FBXW7 Antibody - C-terminal region (ARP47419_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47419_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXW7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human FBXW7. Alternate names: AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59545