Learn More
FBXW7 Mouse anti-Human, Clone: OTI4C11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802883
Catalog No. 50-166-6214
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of maligcies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.
Specifications
Specifications
FBXW7
Monoclonal
Unconjugated
FBXW7
Lyophilized
FBXW7
Human recombit protein fragment corresponding to amino acids 1-354 of human FBXW7 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4C11
PBS with 8% trehalose and no preservative; pH 7.3
Q969H0
AGO, CDC4, FBW6, FBW7, FBX30, FBXO30, FBXW6, SEL-10, SEL10, hAgo, hCdc4
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55294