Learn More
Aviva Systems Biology Corp FBXW8 Antibody - middle region (ARP55585_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBXW8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FBXW8. Alternate names: FBW6, FBW8, FBX29, FBXO29, MGC33534, FBXW6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61546