Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc epsilon RI Armenian Hamster anti-Mouse, DyLight™ 550, Clone: 43160, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143279R

Catalog No. NBP143279R


Add to cart

Description

Description

Fc epsilon RI Monoclonal antibody specifically detects Fc epsilon RI in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications

Specifications

Fc epsilon RI
43160
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Armenian Hamster
IgG
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
FCER1A
The immunogen for this antibody was FceRI alpha.
0.1 mL
Primary
2205
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit