Learn More
Fc epsilon RI Mouse anti-Human, DyLight 755, Clone: 1F2A9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237402IR
Catalog No. NBP237402IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Fc epsilon RI
1F2A9
Western Blot, Flow Cytometry, ELISA
Purified
FCER1A
Purified recombinant fragment of human FCER1A (AA: 42-103) expressed in E. Coli.
Protein A or G purified
Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary
2205
Western Blot, Flow Cytometry, ELISA
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
alpha polypeptide, Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide, Fc-epsilon RI-alpha, FcERI, high affinity immunoglobulin epsilon receptor alpha-subunit, high affinity immunoglobulin epsilon receptor subunit alpha, high-affinity, of mast cells, alpha polypeptide
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human