Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RI/CD64, Human anti-Human, Clone: H22, Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Fc gamma RI/CD64
Clone H22
Applications Flow Cytometry, ELISA
Host Species Human
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015985
View Documents
Novus Biologicals
NBP2810970.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015986
View Documents
Novus Biologicals
NBP2810970.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human Samples. It is validated for Flow Cytometry, Competitive ELISA.
Specifications

Specifications

Fc gamma RI/CD64
Flow Cytometry, ELISA
Human
Primary
H22
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit