Learn More
Fc gamma RI/CD64 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0269- 2Y31, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212175AF594
Catalog No. NP21217A94
Add to cart
Description
Description
Fc gamma RI/CD64 Monoclonal antibody specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Fc gamma RI/CD64
MM0269- 2Y31
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FCGR1A
Human CD64
0.1 mL
Primary
2209