Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc gamma RI/CD64 Rabbit anti-Human, Alexa Fluor 594, Clone: 401, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289362AF594
Description
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry.Specifications
Fc gamma RI/CD64 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2209 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
401 | |
Flow Cytometry | |
CD64 antigen, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FcRI, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fc gamma RI/CD64 (NP_000557.1; Met1-Pro288). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title