Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Fc gamma RIIA/CD32a Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2796635PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rhesus Macaque Fc gamma RIIA/CD32a.Specifications
7.8 pg/mL | |
Conjugated | |
2212 | |
Antibody Pairs | |
7.81-500 pg/mL | |
Storage is content dependent. | |
Mouse | |
Rhesus Macaque | |
FCGR2A | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.5 ng/mL is recommended. |
HRP | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rhesus Macaque Fc gamma RIIA/CD32a. | |
Fc gamma RIIA/CD32a | |
5 Plates | |
Sandwich ELISA | |
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title