Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc gamma RIIA/CD32a Mouse anti-Human, Clone: 3L1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2359360.05MG

Catalog No. NBP23593605


Add to cart

Description

Description

Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Western Blot,Flow Cytometry,CyTOF.
Specifications

Specifications

Fc gamma RIIA/CD32a
3L1
Unconjugated
PBS with no preservative
CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a
Mouse
IgG2
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute the antibody with 500 μL sterile PBS and the final concentration is 200 μg/mL.
CyTOF, Flow Cytometry, Western Blot
LYOPH
Western Blot 1:500 - 1:1000, Flow Cytometry 1:50 - 1:100, CyTOF-ready
P31994
FCGR2A
Human recombinant CD32 EC domain
0.05 mg
Primary
2212
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit