Learn More
Fc gamma RIIIA/CD16a Mouse, DyLight 405, Clone: 3G8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150056V
Catalog No. NBP150056V
Add to cart
Description
Description
Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Antigen in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
Fc gamma RIIIA/CD16a
3G8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
FCGR3A
Human neutrophils.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The mouse monoclonal antibody 3G8 recognizes CD16, a low affinity receptor for aggregated IgG (FcgammaRIII antigen). CD16 exists in two different isoforms: CD16a (FcgammaRIIIA; 50-65 kDa; expressed on NK-cells, monocytes and macrophages) and CD16b (FcgammaRIIIB; 48 kDa; mainly expressed on neutrophils). HLDA V; WS Code NK80
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGRIII, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Mouse
IgG1 Kappa
0.1 ml
Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary
2214
Human, Primate