Learn More
FcεR1α Armenian Hamster anti-Mouse, BUV615, Clone: MAR-1 (also known as MAR1), BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Manufacturer:  BD Biosciences 751770
Catalog No. BDB751770
Add to cart
Description
Description
FcεR1α Monoclonal antibody specifically detects FcεR1α in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
FcεR1α
Flow Cytometry
0.2 mg/mL
Ms FceR1a BUV615 MAR-1 50 μg
Armenian Hamster
1 x 50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Mouse
MAR-1 (also known as MAR1)
Brilliant Ultraviolet 615
BUV615
IgG
RUO
Primary
FceR1a