Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCAMR (CD351) Mouse anti-Human, Mouse, Clone: TX61, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen FCAMR (CD351)
Clone TX61
Concentration 0.5 mg/mL
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry, Functional Assay
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2329100
View Documents
MilliporeSigma
MABF2329100UL
100 μL
Each for $387.00
Add to cart
 
MABF232925
View Documents
MilliporeSigma
MABF232925UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

FCAMR (CD351) Monoclonal antibody specifically detects FCAMR (CD351) in Human, Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry, Affects Function.
Specifications

Specifications

FCAMR (CD351)
0.5 mg/mL
Unconjugated
Mouse
Human, Mouse
Ba/F3 cells transfected with murine FCAMR.
Monoclonal
Protein G purified
TX61
Flow Cytometry, Immunoprecipitation, Immunohistochemistry, Functional Assay
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without azide.
IgG1κ
Primary
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit